Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
$106.25 - $106.25
Specifications
Description Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
Product Type Agar
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R111800
View Documents
Thermo Scientific™
R111800
10/Pk.
Pack of 10 Each for $106.25
Description
Description
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
  • Double-bagged plus a third bag is enclosed for customer use (Double + Bag)
  • Products are manufactured in a clean room and validated sterile by gamma radiation
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Specifications
Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated
Contact - Double Bag
Agar
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Not intended for IVD use.