Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
$205.51 - $644.06
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
|
R20421
|
Thermo Scientific™
R20421 |
Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0) |
Each for $644.06
|
|
|||||
|
R20414
|
Thermo Scientific™
R20414 |
Turbidity Standard 4.0 |
Each for $259.31
|
|
|||||
|
R20412
|
Thermo Scientific™
R20412 |
Turbidity Standard 2.0 |
Each for $267.14
|
|
|||||
|
R20413
|
Thermo Scientific™
R20413 |
Turbidity Standard 3.0 |
Each for $258.23
|
|
|||||
|
R20415
|
Thermo Scientific™
R20415 |
Turbidity Standard 5.0 |
Each for $249.63
|
|
|||||
|
R20410
|
Thermo Scientific™
R20410 |
Turbidity Standard 0.5 |
Each for $205.51
|
|
|||||
|
R20411
|
Thermo Scientific™
R20411 |
Turbidity Standard 1.0 |
Each for $258.03
|
|
|||||
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
- Tube size is 15 x 103mm and fits in most spectrophotometers
- Polystyrene technology allows longer shelf life
Specifications
| 15 x 103 mm |