Learn More
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.
$.00 - $1500.06
Specifications
Control Sets | Positive |
---|---|
Detectable Analytes | beta-haemolytic streptococci |
For Use With (Application) | Enzyme Immunoassay Detection |
Material | Polyvalent |
Organism Group | Group A, B, C, D, F and G |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
DR0704G
|
Thermo Scientific™
DR0704G |
PathoDxtra Strep Group D Latex | 60 Tests |
Pack of 60 Each for $589.66
|
|
||||
DR0701G
|
Thermo Scientific™
DR0701G |
PathoDxtra Strep Group A Latex | 60 Tests |
Pack of 60 Each for $588.86
|
|
||||
DR0707G
|
Thermo Scientific™
DR0707G |
PathoDxtra Polyvalent Positive Control | 2.8 mL |
Each for $391.60
|
|
||||
DR0706G
|
Thermo Scientific™
DR0706G |
PathoDxtra Strep Group G Latex | 60 Tests |
Pack of 60 Each for $589.31
|
|
||||
OXDR0709M
|
Thermo Scientific™
DR0709M |
PathoDxtra Extraction Reagents | 1 Set |
Each for $670.32
|
|
||||
OXDR0700M
|
Thermo Scientific™
DR0700M |
PathoDxtra™ Strep Grouping Kit | 60 Tests/Kit |
Pack of 60 Each for $1,500.06
|
|
||||
DR0720G
|
Thermo Scientific™
DR0720G |
Disposable Reaction Cards | 50 Cards |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
DR0703G
|
Thermo Scientific™
DR0703G |
PathoDxtra Strep Group C Latex | 60 Tests |
Pack of 60 Each for $589.20
|
|
||||
Description
Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.
- Improves workflow efficiencies—instant acid extraction without the need for incubation or reconstitution
- Saves time—simple, rapid procedure produces easy-to-read results
- Reduces costs—identifies all clinically important Strep groups, including Group D; no additional reagents required
- Provides flexibility—pack variants available to suit testing needs
Includes:
PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards
PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit
PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents
Specifications
Positive | |
Enzyme Immunoassay Detection | |
Group A, B, C, D, F and G |
beta-haemolytic streptococci | |
Polyvalent |
Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4