Thermo Scientific™ Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Supplier: Thermo Scientific™ RE30854401
Description
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
2°C to 8°C | |
Yes | |
100 Tests |