Thermo Scientific™ Remel™ QC-Slide™ AFB Stain Control
Remel QC-Slide AFB Stain Control is a heat-fixed quality control slide for acid-fast bacilli stain procedures.
$230.80 - $1218.00
Specifications
Description | AFB Stain Control |
---|---|
Format | Glass Slide |
Product Type | AFB Stain Control |
Description
Remel QC-Slide AFB Stain Control is a heat-fixed quality control slide for acid-fast bacilli stain procedures.
Specifications
AFB Stain Control | |
Glass Slide | |
AFB Stain Control |