Thermo Scientific™ Settling Plate Tryptic Soy Agar
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Tryptic Soy Agar.
$189.26 - $217.26
Specifications
Description Settling Plate Tryptic Soy Agar
Product Type Agar
Quantity 10/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Format Price Quantity  
Catalog Number Mfr. No. Quantity Format Price Quantity  
R111570
View Documents
Thermo Scientific™
RE111570
10/Pk. Settling Monoplate, Single
Pack of 10 Each for $217.26
R111670
missing translation for 'viewDocuments'
Thermo Scientific™
RE111670
10/Pk. Settling Monoplate, Double
Pack of 10 Each for $204.66
R111890
View Documents
Thermo Scientific™
RE111890
10/Pk. 85mm Plate, Double
Pack of 10 Each for $189.26
Description
Description
Isolate and cultivate wide variety of organisms such as bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar. This medium is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. This medium is recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is a general-purpose, nonselective medium providing enough nutrients to allow for a wide variety of microorganisms to grow.5 It is used for a wide range of applications, including culture storage, enumeration of cells (counting), isolation of pure cultures, or simply general culture. Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, USDA3,4
  • Ready-to-use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Settling Plate Tryptic Soy Agar
Agar
10/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Not intended for IVD use.