Thermo Scientific™ Remel™ Tryptic Soy Broth
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
Supplier:  Thermo Scientific™ RE112732
 View more versions of this product
Catalog No. R112732
Explore available promotions
Description
Description
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
Specifications
Specifications
Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
BB Septum - 500mL
10/Pk.
Tryptic Soy Broth
Microbiology Media
10 x 500 mL
Videos
SDS
Documents
Documents
Promotions
Promotions
REF numbers beginning with R11 are not intended for IVD use.