Learn More
Thermo Scientific™ Wellcogen™ Bacterial Antigen Rapid Latex Agglutination Test, Bacterial Meningitis
Rapidly detect bacterial antigens present in human body fluids by latex agglutination.
Supplier: Thermo Scientific™ RE30859602
Description
The Wellcogen™ Bacterial Antigen Kit provides a series of rapid latex tests for use in the qualitative detection of antigen from Streptococcus group B, Haemophilus influenzae type b, Streptococcus pneumoniae (pneumococcus), Neisseria meningitidis (meningococcus) groups A, B, C, Y or W135 and Escherichia coli K1 present in cerebrospinal fluid (CSF) as a consequence of infection. The kit can also be used to test other body fluids or blood culture supernatants for most of these antigens and plate cultures for N. meningitidis group B or Escherichia coli K1. (See Table 1 for indications supported by clinical data). NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; e.g. symptoms, results of other tests, clinical impressions etc.
The kit can also be used to test serum, urine or blood culture supernatants for most of these antigens and plate cultures for N. meningitidis group B or E. coli K1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Streptococcus group B; H. influenzae Type b; S. pneumoniae; N. meningitidis groups A, C, Y, W135; N. meningitidis group B/E. coli K1 |
Wellcogen Bacterial Antigen Kit | |
Antigens in body fluids | |
30 Tests/Kit |