missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ Remote Dispensing System and Accessories for BRAND Dispensette Bottletop Dispensers
Accessories to facilitate remote dispensing with a BRAND Dispensette S or Dispensette S Organic bottletop dispenser. Dispenser not included
$123.53 - $1549.57
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Includes | Price | Quantity | ||||
13-880-710
|
BrandTech™
704261 |
Remote Dispensing System | Dispensing from bulk containers | Tubing, wall mount, 3/4 in. bung adapter |
Each for $1,549.57
|
|
||||
13-880-711
|
BrandTech™
704267 |
FEP Tubing | Remote dispensing | 10 m of tubing |
Each for $1,334.41
|
|
||||
13-880-712
|
BrandTech™
704269 |
FEP Tubing | Remote dispensing | 1 m of tubing |
Each for $123.53
|
|
||||
Description
- 704261- Remote Dispensing System for Dispensette. Includes wall mount
- 704267 - 10 meters of filling tube
- 704269 - 1 meter filling tube
Specifications
Not Autoclavable | |
Non-sterile |