Corning™ Step-R™ Repeating Pipettor Starter Pack
The Corning™ Step-R™ Repeating Pipettor Starter Pack is a convenient, all-in-one kit containing a Repeater dispenser, plus tips in a range of sizes with adapters
Supplier:  Corning™ C6636
Catalog No. 07201491
Explore available promotions
Description
Description
For a smooth start, the Corning™ Step-R™ Repeating Pipettor Starter Pack is packaged with the Step-R Repeating Pipettor, 2 syringe tip adapters (25 mL and 50 mL), and 24 non-sterile syringe tips samples (0.05 mL, 2.5 mL, 5 mL, 12.5 mL, 25 mL, and 50 mL).
  • Easy volume setting: Volume selection chart conveniently placed on the handle
  • Fast and precise multiple dispensing: Up to 48 pipetting doses without refilling the tip syringe
  • Maintenance-free
  • Meets ISO 8655 requirements
  • Delivered with QC certificate
Learn more from North America
Learn more from Europe, Middle East, and Africa
See resource tab below for specifications.
'
Specifications
Specifications
ISO8655
Non-sterile
0.05, 2.5, 5, 12.5, 25, 50 mL
1/Pk.
Autoclavable
Repeating Pipette Starter Pack
Dispensing of viscous and volatile liquids