missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glastron Repeating Dispensers DFS Item

Fixed-volume models from 1 through 100mL

Supplier:  Glastron TG50360005

 View more versions of this product

Catalog No. 13-683-10B


Explore available promotions

Description

Description

  • Consist of borosilicate flask with standard taper 24/40 joint and interchangeable pipet head mounted on top
  • Simply tilt back flask to fill pipet to designated volume
  • Volume is dispensed when flask is tilted forward
  • Accurate to approximately ±2%

Specifications

Specifications

5 mL
Interchangeable pipet head
Borosilicate Glass
24/40
Videos
SDS
Documents

Documents

Promotions

Promotions