missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FEATHER Replaceable Blade Scissors No.145

FEATHER Replaceable Blade Scissors No.145 includes a handle that can be used for all three different blade tip types: sharp/sharp, sharp/blunt and blunt/blunt and 1 set of replacable blades. The replaceable blade design eliminates the cost and trouble of resharpening.

$117.26 - $138.61

Specifications

Product Type Scissors with Replacable Blades
Length (Metric) 132 mm
Length (Metric) Blade 48 mm
Material Stainless Steel
Autoclavable Autoclavable
View More Specs

Products 3
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
13-804-27
View Documents
FEATHER
145SHBL
Sharp/Blunt
Each for $138.61
 
361042803
View Documents
FEATHER
145SHSH
Sharp/Sharp
Each for $117.26
 
361042804
View Documents
FEATHER
145BLBL
Blunt/Blunt
Each for $121.13
 
Description

Description

  • Made in Japan
  • Sharp/Sharp or Sharp/Blunt or Blunt/Blunt styles available
  • Stainless steel handle and blade
Specifications

Specifications

Scissors with Replacable Blades
48 mm
Autoclavable
132 mm
Stainless Steel
SDS
missing translation for 'documents'

missing translation for 'documents'