Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector
Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Supplier:  Fisherbrand™ LDAGI104LLFB
 View more versions of this product
Catalog No. 14-500-720
Explore available promotions
Description
Description
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Detector Lamp
Deuterium
Item 14-500-720 is comparable to Agilent part number G1314-60100.
Agilent 1100, 1200, 1260 UV and Visible HPLC Detector