Thermo Scientific™ Replacement Microcentrifuge Rotor Lids
Combine these replacement microcentrifuge rotor lids with the microcentrifuge rotor.
$822.23 - $893.96
Products 2
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
05-375-108
View Documents
Thermo Scientific™
75003406
Screw-on Lid
Each for $822.23
05-375-107
View Documents
Thermo Scientific™
75003410
ClickSeal™ Biocontainment Lid
Each for $893.96
Description
Description
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
  • For use with 24 × 1.5/2.0 mL Rotor (75003424), PCR 4 × 8 Rotor (75003440)
  • Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
  • With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
  • For use with 36 × 0.5 mL Rotor (75003436), Dual Row 18 × 2.00.5 mL Rotor (75003418), PCR 8 × 8 Rotor (75003489)
Ordering Information:
Sold individually.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'