Thermo Scientific™ Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Supplier:  Thermo Scientific™ 365RN235
 View more versions of this product
Catalog No. 36-5RN-235
Explore available promotions
Description
Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Specifications
Specifications
365RN235
1.9685 in.
1 mL
Bevel
Removable
23 G
Replacement Syringe Needle
50 mm
Thermo 365K3041 Syringe
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.