Hamilton™ Replacement Needles for Waters U6K HPLC Injection Valve Syringes
Small and large hub removable replacement needles
Supplier:  Hamilton™ HAM90049
 View more versions of this product
Catalog No. 14-815-426
Explore available promotions
Description
Description
Needles are available as a 25s gauge with a small hub for syringes 2.5-100οL in volume, a large hub for syringes 250οL and larger, and metal hub needles for TLL syringes.
Six needles per pack.
Specifications
Specifications
Autoclavable
25s G
Metal Hub Replacement Needle
Waters U6K HPLC Injection Valve Syringes
Metal (Hub)
1.97 in.
Square/Blunt