missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bradley™ Barrier-Free Vertical-Supply Drench Shower Accessory, Ball Valve

Durable brass fixture for use with drench shower.

Supplier:  Bradley™ S27278

Catalog No. 19-807-556


Explore available promotions

Description

Description

  • Chrome plated brass for extra durability
  • Silver color
Specifications

Specifications

Silver
3.06 in.
Chrome-plated Brass
1.1 lb.
Drench Shower
7.7 cm
Ball Valve
0.49 kg
Videos
SDS
Documents

Documents

Promotions

Promotions