Synthware™ Replacement Straight Bore Stopcock
Supplied with PTFE plug, one PTFE washer and one O-ring.
Supplier: Synthware™ S181125
Specifications
Straight Bore Stopcock | |
Borosilicate Glass | |
Straight Bore Tap | |
PTFE |
2 mm | |
Organic Synthesis | |
PTFE plug, One PTFE washer, One o-ring |