Brady™ Repositionable Vinyl Cloth Wire and Cable Labels
Brady™ Repositionable Vinyl Cloth Wire and Cable Labels features CleanLift™ B-498 Series Vinyl Cloth with strong permanent adhesive that is also repositionable if needed.
Supplier: Brady™ M7498
Description
- The adhesive and cloth backing offers excellent holding power, yet allows for clean removal and repositioning
- Sticks to any clean, dry surface
- Specially formulated topcoat for excellent print quality
- All-in-one cartridge includes label supply and R6200 Series ribbon
Specifications
Label | |
BMP41 Label Printer, BMP51 Label Printer, BMP53 Label Printer | |
Thermal Transfer | |
Non-sterile | |
Cartridge, Ribbon | |
240 Pk. |
Black on White | |
Vinyl Cloth (B-498) | |
0.5 x 0.5 in. | |
Semi-gloss | |
UL Recognized to UL969, RoHS Compliant to 2005/618/EC MCV |