Eppendorf™ Research™ plus Move It™, Mechanical Multi-Channel Pipettes with Adjustable Tip Spacing
Allows easy format changes with 70% time-savings compared to single-channel pipettes. The unique, tubeless design is autoclavable to reduce the risk of contamination and improve sample integrity.
$3700.84 - $5692.50
Specifications
Certifications/Compliance Lot-Specific Certificate
Compatible Tips Universal Tips, epT.I.P.S.™, epT.I.P.S.™ 384
Adjustable Tip Spacing 4.5 to 33 mm
Autoclavable Autoclavable
For Use With Air-Cushion
View More Specs
Products 9
Catalog Number Mfr. No. Number of Channels Volume (Metric) Includes Color Price Quantity  
Catalog Number Mfr. No. Number of Channels Volume (Metric) Includes Color Price Quantity  
05414056
View Documents
Eppendorf™
3125000150
4 30 to 300 μL epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips Orange
Each for $5,289.05
05414059
View Documents
Eppendorf™
3125000184
4 120 to 1200 μL Without Box Dark Green
Each for $5,289.05
05414057
View Documents
Eppendorf™
3125000168
6 30 to 300 μL epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips Orange
Each for $5,511.21
05414060
View Documents
Eppendorf™
3125000192
6 120 to 1200 μL Without Box Dark Green
Each for $5,511.21
361057842
View Documents
Eppendorf™
3125000117
8 1 to 20 μL epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips Light Pink
Each for $3,700.84
01-670-721
View Documents
Eppendorf™
3125000133
8 5 to 100 μL epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips Light Yellow
Each for $3,700.84
05414058
View Documents
Eppendorf™
3125000176
8 30 to 300 μL epT.I.P.S.™Â Box 2.0 with 96 Pipette Tips Orange
Each for $5,692.50
05414061
View Documents
Eppendorf™
3125000206
8 120 to 1200 μL Without Box Dark Green
Each for $5,692.50
01-670-717
View Documents
Eppendorf™
3125000141
12 5 to 100 μL epT.I.P.S.™Â Box 2.0 with 384 Pipette Tips Light Yellow
Each for $3,963.12
Description
Description
  • Air-cushion pipettes with variable volume and adjustable tip spacing are available in 4, 6, 8 and 12-channel
  • Choose from Mechanical (Research™ plus) or Electronic (Xplorer™ plus) adjustable tip spacing multichannel pipettes
  • For Plates (12, 24, 48, 96, and 384-well), Tubes (1.5 and 2 mL), and Agarose Gels
  • Tip spacing is freely selectable from 4.5 to 33 mm, depending on the pipette variant
  • Spacing control for quick switches backwards and forwards between source and destination formats
  • Manual adjustment knob for smooth, vibration-free format switches and drip-free liquid transfers
  • 360° rotatable pipette head for comfortable readability of display and fast parameter identification
  • Tubeless system is safe and reliable
  • Optimally balanced pipette reduces hand fatigue for stress-free operation
  • Autoclavable for user and sample safety (Research™ plus fully, and Xplorer™ plus lower part only)
Specifications
Specifications
Lot-Specific Certificate
4.5 to 33 mm
Air-Cushion
Mechanical Pipette
Universal Tips, epT.I.P.S.™, epT.I.P.S.™ 384
Autoclavable
Research™ Plus™
Air-Cushion
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'