MilliporeSigma™ Supelco™ Reservoir for Filtration Apparatus 1
Replacement glass reservoir for Supelco™ mobile phase filtration apparatus 1
$217.92 - $273.36
Specifications
For Use With (Application) | Liquid Chromatography |
---|---|
For Use With (Equipment) | Filtration Apparatus 1 |
Description
Reservoir for Filtration Apparatus 1 protects the instrument and columns by removing particles and gases from solvents and other mobile phase components.Specifications
Liquid Chromatography | |
Filtration Apparatus 1 |