missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Resorcinol, 99%, Thermo Scientific Chemicals

$40.59 - $1150.08

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1308018
View Documents
Thermo Scientific Chemicals
A1308018
50 g
Each for $40.59
 
AAA1308030
View Documents
Thermo Scientific Chemicals
A1308030
250 g
Each for $79.03
 
AAA130800B
View Documents
Thermo Scientific Chemicals
A130800B
1000 g
Each for $271.90
 
AAA130800I
View Documents
Thermo Scientific Chemicals
A130800I
5000 g
Each for $1,150.08
 
Description

Description

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

Solubility
Soluble in water, alcohol, ether and glycerol.

Notes
Stable. May discolor on exposure to air or light.
Specifications

Chemical Identifiers

108-46-3
110.112
GHMLBKRAJCXXBS-UHFFFAOYSA-N
5054
benzene-1,3-diol
C6H6O2
MFCD00002269
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
CHEBI:27810
C1=CC(=CC(=C1)O)O

Specifications

108-46-3
1.272
280°C to 281°C
Phenol-like
MFCD00002269
UN2876
Air and light sensitive
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
5054
110.11
Resorcinol
109°C to 112°C
4 to 6
171°C (339°F)
C6H6O2
50 g
906905
14,8155
Soluble in water,alcohol,ether and glycerol.
C1=CC(=CC(=C1)O)O
110.112
CHEBI:27810
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H301-H315-H319-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: RESORCINOL

EINECSNumber : 203-585-2

RTECSNumber : VG9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only