missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Resorcinol, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03624836

 View more versions of this product

Catalog No. AA3624836


Explore available promotions

Description

Description

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Resorcinol
108-46-3
White to Cream
4 to 6
171°C (339°F)
Phenol-like
MFCD00002269
UN2876
Air and light sensitive
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
5054
110.11
ACS
For analysis ACS
109°C to 112°C
1.272 g/mL
280°C to 281°C
99.0 to 100.5%
C6H6O2
500 g
906905
14,8155
Soluble in water,alcohol,ether and glycerol.
C1=CC(=CC(=C1)O)O
110.112
CHEBI:27810
99.0 to 100.5%
Crystalline

Chemical Identifiers

108-46-3
110.112
GHMLBKRAJCXXBS-UHFFFAOYSA-N
5054
benzene-1,3-diol
C6H6O2
MFCD00002269
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
CHEBI:27810
C1=CC(=CC(=C1)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H301-H315-H319-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: RESORCINOL

EINECSNumber : 203-585-2

RTECSNumber : VG9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only