Dentec Safety™ Respirator Wipes
Conveniently packaged wipes clean and disinfect respirators and other PPE equipment.
Supplier:  Dentec Safety™ DTC12A311
 View more versions of this product
Catalog No. 19222536
Explore available promotions
Description
Description
  • Small packages can be stored anywhere - pocket, purse, lunch boxes, etc
  • Safe and effective for cleaning respirators, hard hat suspensions, and other personal protective equipment
  • Also useful for cleaning phones and keyboards
  • Measures 5 x 8 inches when unfolded
Specifications
Specifications
Respirator Wipes
Alcohol-free
10 Boxes; 100/Box
79100
Facepieces
5 x 8 in. (unfolded)