Velp Scientifica™ RESPIROMETRIC Maxi without Databox
The versatile, reliable and cloud-enabled solution designed for anaerobic respiration studies such as the Biochemical Methane Potential (BMP) of biomass, without the databox.
Supplier:  Velp Scientifica™ SB10210156
Catalog No. 07222021
Explore available promotions
Description
Description
  • Designed to support anaerobic respiration studies to determine the Biochemical Methane Potential (BMP) of biomass
  • BMP analysis studies the production of biogas from organic wastes mainly for Waste and Wastewater treatment and for Renewable Energy Production
  • Bright 4-digit LED display ensures long battery life
  • Intuitive 2 button interface to facilitate the analysis set up
  • Pressure range: 500/2000 mbar (hPa)
  • Results available in BMP (NmLCH4/gSV) and Pressure (mbar)
  • Sensor has the exclusive VELP™ Ermes Cloud Platform
Specifications
Specifications
Sensor System
IP54
CE