Velp Scientifica™ RESPIROMETRIC Sys6 without Databox
The compact, reliable, cloud-enabled solution for BOD determination and aerobic respirometric tests.
Supplier: Velp Scientifica™ SB10210146
Description
- Designed to provide a smart and cloud-enabled solution to laboratories of water and wastewater quality control
- BOD (Biochemical Oxygen Demand) Determination is a simple and easy way for monitoring the microorganisms' activity in water samples with extremely reliable results
- Microorganisms present or inoculated in a water sample containing biodegradable organic substance, consume oxygen for their metabolic activity and produce a corresponding volume of carbon dioxide
- Carbon dioxide gets absorbed by a strong alkali positioned in the neck of the bottle
- Result of this process is a decrease in gas pressure
- BOD measures this change in gas pressure between the beginning and the end of the analysis and gives a result expressed in mg/L
Specifications
Sensor System | |
IP54 | |
CE |