Learn More
Restek™ 11mm GC Snap-Top Cap with Septum DFS Item
An effecitve alternative when there is no need to crimp caps
$108.32 - $1135.81
Specifications
Color Clear
Material Polypropylene
Closure Size 11 mm
View More Specs
Products 6
Catalog Number Mfr. No. Quantity Material (Liner) Price Quantity  
Catalog Number Mfr. No. Quantity Material (Liner) Price Quantity  
06718624 DFS Item
View Documents
Restek™
RK21729
100/Pk. PTFE
Pack of 100 Each for $108.32
Only null left
06718625 DFS Item
View Documents
Restek™
RK21730
1000/Pk. PTFE
Pack of 1000 Each for $734.77
Only null left
06718627 DFS Item
View Documents
Restek™
RK21732
1000/Pk. PTFE/Butyl Rubber
Pack of 1000 Each for $896.42
Only null left
06718628 DFS Item
View Documents
Restek™
RK21733
100/Pk. PTFE/Silicone
Pack of 100 Each for $303.94
Only null left
06718626 DFS Item
View Documents
Restek™
RK21731
100/Pk. PTFE/Butyl Rubber
Pack of 100 Each for $129.15
Only null left
06718629 DFS Item
View Documents
Restek™
RK21734
1000/Pk. PTFE/Silicone
Pack of 1000 Each for $1,135.81
Only null left
Description
Description
  • Snaps on any crimp-style vial
  • Clear
  • Polypropylene
Specifications
Specifications
Clear
Polypropylene
11 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'