Restek™ 2-Inch Vacuum Gauge DFS Item
High-quality vacuum gauges with 316 stainless steel wetted surfaces
Supplier:  Restek™ RK24269
 View more versions of this product
Catalog No. 06712267
Explore available promotions
Description
Description
It is recommended for use with passive sampling kits.
  • -30 in. Hg to 0 in. Hg
  • Rear mount
  • Diameter: 2 in.
Specifications
Specifications
Stainless Steel wetted surfaces
Vacuum Gauge
1/8 in.
2 in.
For use with passive sampling kits
Analog