Restek™ Dual Vespel™ Ring Inlet Seals DFS Item
Extend column lifetime by preventing oxygen from reaching the column with these inlet seals. Restek™ Dual Vespel™ Ring Inlet Seals eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.
Supplier:  Restek™ RK21248
Catalog No. 06-802-810
Explore available promotions
Description
Description
  • Washerless, Leak-Tight 1.2mm Seals for Agilent GCs
  • Two per pack
  • Does not require retightening of reducing nut after several oven cycles
  • Same price as the regular inlet seals with washers
  • Siltek treatment provides enhanced inertness versus stainless steel
  • Our patented dual Vespel ring inlet seal greatly improves injection port performance
  • It stays sealed, even after repeated temperature cycles, without retightening the reducing nut
  • This seal features two soft Vespel rings, one embedded in its top surface and the other embedded in its bottom surface
  • The rings will not harm the critical seal in the injector body, or any other surface, and are outside the sample flow path
  • Use a stainless steel seal for analyses of unreactive compounds
  • To reduce breakdown and adsorption of active compounds, use a gold-plated or Siltek-treated seal
Specifications
Specifications
1.2 mm
Siltek-treated