missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Restek™ Glass Vial Inserts
Specially designed for 2 mL, 11 mm crimp top and 2 mL, 9 mm short cap, screw thread vials
Supplier: Restek™ RK21779
Description
- Limited-volume inserts also available
- Material: glass
Specifications
Used with 2.0 mL, 11 mm Crimp-Top Vials; 2.0 mL, 10 mm Big Mouth Step Design Screw-Thread Vials | |
250 μL | |
Glass Flange (step) |
Glass | |
1000 | |
Big Mouth Step Design Vials |