Restek™ Glass Vial Inserts
Specially designed for 2 mL, 11 mm crimp top and 2 mL, 9 mm short cap, screw thread vials
Supplier: Restek™ RK24510
Description
- Limited-volume inserts also available
- Material: glass
Specifications
Glass | |
1000 | |
Standard Mouth Vials |
250 μL | |
Flat Bottom |