missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Restek™ Headspace 20mm Autosampler Vials
For use in the concentration and analysis of volatile organic compounds
$152.36 - $2351.19
Specifications
Color | Clear |
---|---|
For Use With (Application) | Concentration and analysis of volatile organic compounds |
Closure Type | Crimp Cap |
Closure Size | 20 mm |
Catalog Number | Mfr. No. | Volume (Metric) | Base Style | Quantity | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Base Style | Quantity | Price | Quantity | |||
06718455
![]() |
Restek™
RK21160 |
27 mL | Flat bottom | 100 Pk. |
Pack of 100 Each for $295.02
|
|
|||
06718456
![]() |
Restek™
RK21161 |
27 mL | Flat bottom | 1000 Pk. |
Pack of 1000 Each for $2,351.19
|
|
|||
06718457
![]() |
Restek™
RK21162 |
20 mL | Round bottom | 100 Pk. |
Pack of 100 Each for $190.40
|
|
|||
06718459
![]() |
Restek™
RK21164 |
10 mL | Round bottom | 100 Pk. |
Pack of 100 Each for $184.70
|
|
|||
06718458
![]() |
Restek™
RK21163 |
20 mL | Round bottom | 1000 Pk. |
Pack of 1000 Each for $1,389.34
|
|
|||
06718461
![]() |
Restek™
RK21165 |
10 mL | Round bottom | 1000 Pk. |
Pack of 1000 Each for $1,345.34
|
|
|||
06718463
![]() |
Restek™
RK21166 |
6 mL | Flat bottom | 100 Pk. |
Pack of 100 Each for $152.36
|
|
|||
06718464
![]() |
Restek™
RK21167 |
6 mL | Flat bottom | 1000 Pk. |
Pack of 1000 Each for $1,088.87
|
|
|||
06719016
![]() |
Restek™
RK24683 |
10 mL | Flat bottom | 100 Pk. |
Pack of 1000 Each for $155.94
|
|
|||
06719017
![]() |
Restek™
RK24684 |
10 mL | Flat bottom | 1000 Pk. |
Pack of 1000 Each for $1,322.85
|
|
|||
06719019
![]() |
Restek™
RK24686 |
20 mL | Flat bottom | 1000 Pk. |
Pack of 1000 Each for $1,420.66
|
|
|||
06719018
![]() |
Restek™
RK24685 |
20 mL | Flat bottom | 100 Pk. |
Pack of 100 Each for $207.85
|
|
|||
Description
- Compatible with numerous instruments
- Color: Clear
Specifications
Clear | |
Concentration and analysis of volatile organic compounds | |
Crimp Cap | |
20 mm |