Restek™ HPLC 30-Column Storage Cabinet
Saves space and protects columns.
Supplier: Restek™ RK25159
Specifications
Hinged door | |
Hinged door is clear for quick identification of column labels or tags | |
Easy-to-install cabinet saves space and protects columns, the hinged door is clear to allow quick identification of column labels or tags | |
Easy-to-install, wall-mountable (screws not included) | |
PVC |
HPLC column storage Cabinet | |
17 x 15 x 3 in. | |
HPLC columns (min. 100mm L) | |
30 columns (minimum 100 mm L) |