Restek™ Inlet Liners for Splitless Injection - Cyclo Double Gooseneck DFS Item
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Supplier:  Restek™ RK20896
 View more versions of this product
Catalog No. 06718140
Explore available promotions
Description
Description
  • Compatible with Agilent™/Finnigan columns
  • Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples
  • To order liners in pack quantities, please contact your Fisher Scientific™ Customer Service Representative
Specifications
Specifications
IP Cyclo Double Taper Gooseneck Inlet Liner
7.85 cm
0.65 cm
Borosilicate Glass
5/Pk.
0.4 cm
Agilent GCs Equipped with Split/Splitless Inlets