Restek™ Instrument-Grade Stainless Steel Tubing
Used for providing carrier, fuel, make-up, or auxiliary gases to laboratory instruments.
Supplier: Restek™ RK21500
Description
- Material: Instrument-grade 304 grade stainless steel
Order Info
Maximum continuous lengths are: 2000 ft. (1/16 in. OD) and 750 ft. (1/4 in. OD). The availability of long lengths is subject to inventory constraints. Lead times may vary depending on the continuous length needed. Please inquire before ordering.
Specifications
Welded/Drawn Tubing | |
0.01 in. | |
0.158 cm | |
7.62 m |
For Providing Carrier, Fuel, Make-up or Auxiliary Gases to Laboratory Instruments | |
0.0625 in. | |
25 ft. | |
Stainless Steel (304) |