Restek™ Mininert Precision Sampling Valves for Vials DFS Item
Convenient valves for repetitive sampling and limiting content exposure to the silicon septum.
Supplier:  Restek™ RK24904
 View more versions of this product
Catalog No. 06719054
Explore available promotions
Description
Description
  • Available for both screw-cap and crimp-top vials
  • Crimp-top valve for 20mm ID glassware slides into the neck of the vial
  • Turn the threaded flange to secure a tight fit
Specifications
Specifications
Valve
12 Pack
Screw-Thread Vials
Precision Sampling
24 mm/400