Restek™ MXT-1 Silcosteel Capillary Columns - 60m (197 ft.) Length DFS Item
Siltek™-treated stainless steel
Supplier:  Restek™ RK70143
 View more versions of this product
Catalog No. 06713375
Explore available promotions
Description
Description
  • Nonpolar phase; Crossbond™ dimethyl polysiloxane
  • General-purpose columns for solvent impurities, PCB congeners (e.g., Aroclor mixes), gases, natural gas odorants, sulfur compounds, essential oils, hydrocarbons, semivolatiles, pesticides, and oxygenate
  • Temperature range: -60°C to +430°C (-74° to +806°F)
  • Equivalent to USP G1, G2, G38 phases
Specifications
Specifications
0.53 mm
Mxt-1
Essential Oils Analysis, PCB Congener Analysis, Gases Analysis, Natural Gas Odorants Analysis, Sulfur Compounds Analysis, Essential Oils, Hydrocarbon Analysis, Semivolatiles Analysis, Pesticide Analysis, Oxygenates Analysis
-60°C to +400°C
60 m
Non polar
G1, G2, G38
445°C
0.5 μm
0.5 μm
Stainless Steel
Siltek™