Restek™ Polypropylene Vial Inserts
Specially designed for 2.0mL, 11mm crimp-top and 2.0mL, 9mm short-cap, screw-thread vials.
$63.02 - $145.87
Catalog Number | Mfr. No. | Volume (Metric) | Type | Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Type | Style | Price | Quantity | |||
06718927
|
Restek™
RK24511 |
100 μL | Vial Insert | Big Mouth without Bottom Spring |
Pack of 100 Each for $93.83
|
|
|||
06718933
|
Restek™
RK24518 |
250 μL | Vial Insert | Big Mouth with Bottom Spring |
Pack of 100 Each for $145.87
|
|
|||
06718935
|
Restek™
RK24520 |
250 μL | Vial Insert | Big Mouth without Spring |
Pack of 100 Each for $63.02
|
|
|||
Description
- Big mouth
- Pack of 100
Specifications
Polypropylene | |
100/Pk. | |
Vial Insert |