Restek™ Replacement Combination Vacuum/Pressure Gauges
2 in. vacuum/pressure gauges
Supplier: Restek™ RK24104
Description
- 316 stainless steel
- 1/8 in. NPT fitting
- Center back mount
Specifications
Vacuum/Pressure Gauge | |
30 in. to 30 psig | |
316 Stainless Steel | |
For canisters |