Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments DFS Item
Replacement weldments for Agilent 5890 and 6890/6850 GCs
Supplier:  Restek™ RK20266
 View more versions of this product
Catalog No. 06710553
Explore available promotions
Description
Description
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
Specifications
Gas Chromatography
Agilent 5890 GCs
Stainless Steel
Shell Weldment