Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Replacement weldments for Agilent 5890 and 6890/6850 GCs
Supplier: Restek™ RK20267
Description
- Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
- High-quality stainless steel injector
- Meets or exceeds Agilent original equipment performance
Specifications
Gas Chromatography | |
Manual Flow or Epc on Agilent 5890 GCs; Manual Flow Only on Agilent 6890/6850 GCs | |
Siltek Treated Stainless Steel | |
Weldment |