Learn More
Restek™ Rtx™-1701 Capillary Columns DFS Item
General-Purpose Columns for Alcohols, Oxygenates, PCB Congeners (e.g., Aroclor Mixes), Pesticides, and Fragrance Compounds
$1630.08 - $1949.06
Specifications
Stationary Phase Rtx™-1701
USP Type G46
Functionality/Interaction Type Cyano and Phenyl
Coating Fused Silica
View More Specs
Products 7
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity  
06709620 DFS Item
View Documents
Restek™
RK12070
30 m 0.53 mm 1.5 μm
Each for $1,949.06
Only null left
06709597 DFS Item
View Documents
Restek™
RK12055
30 m 0.53 mm 1 μm
Each for $1,949.06
Only null left
06709591 DFS Item
View Documents
Restek™
RK12053
30 m 0.25 mm 1 μm
Each for $1,630.08
Only null left
06709541 DFS Item
View Documents
Restek™
RK12024
30 m 0.32 mm 0.25 μm
Each for $1,744.47
Only null left
06709568 DFS Item
View Documents
Restek™
RK12039
30 m 0.32 mm 0.5 μm
Each for $1,831.87
Only null left
06709534 DFS Item
View Documents
Restek™
RK12023
30 m 0.25 mm 0.25 μm
Each for $1,630.08
Only null left
06709594 DFS Item
View Documents
Restek™
RK12054
30 m 0.32 mm 1 μm
Each for $1,744.47
Only null left
Description
Description
  • Mix of cyano and phenyl functional groups increase polarity
  • Offers a different elution order relative to less polar Rtx™-1 or Rtx™-5 columns
  • Ideal for confirmation analysis in combination with an Rtx™-35 or Rtx™-5 column
  • Polymer is fully characterized to ensure long-term reproducibility, column-to-column consistency, and low bleed
  • For Alcohols, Oxygenates, PCB Congeners (e.g., Aroclor Mixes), Pesticides, and Fragrance Compounds
Specifications
Specifications
Rtx™-1701
G46
Cyano and Phenyl
Fused Silica
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'