Restek™ Rtx™-VMS Capillary Columns: Standard DFS Item
Provide complete separation of all U.S. EPA Method 8260 compounds in less than 18 minutes
Supplier:  Restek™ RK49915
 View more versions of this product
Catalog No. 06713261
Explore available promotions
Description
Description
Columns only.
  • Special polymer formulation features low bleed, improved selectivity, and faster analysis time for separation of volatile organic compounds by GC/MS
  • Excellent thermal stability
  • Individual test chromatogram is provided with each column
Specifications
Specifications
0.18 mm
350°C
1 μm
1 μm
Stainless Steel
Rtx™-V MS
Volatile Organic Pollutants Analysis
-40°C to +240/260°C
40 m
Fused Silica