Restek™ Soap Film Bubble Flowmeter DFS Item
For flows between 0.1 and 10 cc/min. requires 1mL flowmeter
Supplier:  Restek™ RK20135
 View more versions of this product
Catalog No. 06710511
Explore available promotions
Includes: A reservoir bulb, 24 in. (60cm) of 1/4 in. ID tubing, adapter tubes for 1/8 in. tubing and 0.53mm ID capillary columns, and Velcro™ fasteners.
Description
Description
  • 50mL flowmeter for flows between 10 and 300cc/min.
Specifications
Specifications
Bubble Flowmeter
0.1 and 10 cc/min.
0.03 oz.
1 mL
Product Suggestions
Product Suggestions