Restek™ Clean-Cut Tubing Cutter
Cuts polymeric tubing up to 1/8 in. OD
Supplier: Restek™ RK25069
Description
- Burr-free, perpendicular cuts that will not distort the tubing OD or close the ID
- Use on PEEK, PTFE, Tefzel, and other polymeric tubing
Specifications
Tubing Cutter | |
HPLC | |
1 Each | |
For PEEK in., Teflon in., Tefzel in., Other Polymeric Tubing |