Restek™ Syringe Adapter Kit DFS Item
For Single-Stage VOC Regulator
Supplier:  Restek™ RK21118
 View more versions of this product
Catalog No. 06711441
Explore available promotions
Includes: One nickel-plated brass 1/4 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, One stainless steel, 1/4 in. NPT x 1/8 in. compression fitting with septum (can be used with any syringe needle).
Description
Description
  • Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator
Specifications
Specifications
Syringe Adaptor Kit
Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator
VOC single stage regulators
One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle)