Restek™ Universal 10-32 PEEK Column Connector DFS Item
Universal PEEK connectors allow easy installation of all 0.0625 in. tubing, including stainless steel, up to 5000psi.
$198.77 - $266.94
Products 2
Catalog Number Mfr. No. Color For Use With Price Quantity  
Catalog Number Mfr. No. Color For Use With Price Quantity  
06719061 DFS Item
View Documents
Restek™
RK25016
Black For Universal 10-32 PEEK Column Plμg
Pack of 10 Each for $198.77
06719060 DFS Item
View Documents
Restek™
RK25015
Beige For 1/16 in. Tubing
Pack of 10 Each for $266.94
Description
Description
  • Quantity: 10/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'