Learn More
Promega Restriction Enzymes - EcoR V Blue/White Cloning Qualified (D)
Capable of digesting DNA in 15 minutes or less. (GAT/ATC CTA\TAG)
Supplier: Promega R6355
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Specifications
| 10 U/μL | |
| 37°C | |
| Capable of digesting DNA in <15 min. | |
| -30°C to -10°C | |
| GAT.ATC | |
| Liquid |
| 1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U EcoRV (HC), 1 x 0.25mL Multi-Core™ 10X Buffer | |
| 7.4 | |
| 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
| 10,000 U | |
| EcoRV |