missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BUCHI Retaining Container for Solvent Bottle Platform

Retaining Container for solvent bottle platform provides additional safety in regards to leaking.

Supplier:  BUCHI 11068468

Catalog No. 05405494


Explore available promotions

Description

Description

  • For use with BUCHI Pure Chromatography Systems
Specifications

Specifications

Container
Leak Prevention
Solvent Bottle Platforms
Videos
SDS
Documents

Documents

Promotions

Promotions