Learn More
RETSCH Drying Container
Used for Fluid Bed Dryer TG 200.
Supplier: RETSCH 727830002
Description
- Used for Fluid Bed Dryer TG 200
Specifications
| Drying Container | |
| 6 L | |
| Stainless Steel | |
| Fluid Bed Dryer TG 200 |